Search
Search
| Gene Symbol: Cdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053393.2 | NP_445845.2 | 6 - 7 | 1559 | 78 | |
| XM_008772244.1 | XP_008770466.1 | 5 - 6 | 906 | 78 | ||
| GenBank mRNA | AB010436.1 | - | 6 - 7 | 1559 | 78 | |
| AB010437.1 | - | 6 - 7 | 1559 | 78 | ||
Set Membership: |
|