Search
Search
| Gene Symbol: Cnot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134840.1 | NP_001128312.1 | 29 - 30 | 4312 | 61 | |
| XM_008772271.1 | XP_008770493.1 | 29 - 30 | 4927 | 61 | ||
| XM_008772272.1 | XP_008770494.1 | 29 - 30 | 4912 | 61 | ||
Set Membership: |
|