Search
Search
| Gene Symbol: Mrps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109518.1 | NP_001102988.1 | 1 - 2 | 102 | 70 | |
| GenBank mRNA | FQ210083.1 | - | 1 - 2 | 57 | 70 | |
| FQ228795.1 | - | 1 - 2 | 80 | 70 | ||
Set Membership: |
|