Search
Search
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130428.1 | NP_569112.1 | 11 - 12 | 1570 | 114 | |
| GenBank mRNA | AB072907.1 | - | 11 - 12 | 1570 | 114 | |
| DQ402976.1 | - | 9 - 10 | 1379 | 114 | ||
Set Membership: |
|