Search
Search
| Gene Symbol: Psmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198730.2 | NP_942025.1 | 2 - 3 | 396 | 85 | |
| GenBank mRNA | BC059159.1 | - | 2 - 3 | 401 | 6887177 | 85 |
| FQ212955.1 | - | 2 - 3 | 396 | 85 | ||
| FQ213627.1 | - | 2 - 3 | 420 | 85 | ||
| FQ214459.1 | - | 2 - 3 | 396 | 85 | ||
| FQ215751.1 | - | 2 - 3 | 396 | 85 | ||
| FQ229068.1 | - | 2 - 3 | 413 | 85 | ||
Set Membership: |
|