Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01538446_m1
          See other ARHGAP12 GE Assays ›
          Gene Symbol:
          Arhgap12
          Gene Name:
          Rho GTPase activating protein 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 54280817 - 54394716 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01538446_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgap12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107357.1 NP_001100827.1 18 - 19 2600 95
          XM_006254011.2 XP_006254073.1 18 - 19 2646 95
          XM_006254012.2 XP_006254074.1 17 - 18 2571 95
          XM_006254013.2 XP_006254075.1 17 - 18 2506 95
          XM_008771727.1 XP_008769949.1 16 - 17 2431 95
          XM_008771728.1 XP_008769950.1 15 - 16 1676 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000784.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s)

          morphogenesis of an epithelial sheet
          signal transduction

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68830 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174506 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100530 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100530 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4911 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13210 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795577 - -
          Affymetrix Rat Gene 1.0 ST Array 10795594 - -
          Affymetrix Rat Gene 1.0 ST Array 10795576 - -
          Affymetrix Rat Gene 1.0 ST Array 10795589 - -
          Affymetrix Rat Gene 1.0 ST Array 10795590 - -
          Affymetrix Rat Gene 1.0 ST Array 10795588 - -
          Affymetrix Rat Gene 1.0 ST Array 10795592 - -
          Affymetrix Rat Gene 1.0 ST Array 10795587 - -
          Affymetrix Rat Gene 1.0 ST Array 10795582 - -
          Affymetrix Rat Gene 1.0 ST Array 10795584 - -
          Affymetrix Rat Gene 1.0 ST Array 10795586 - -
          Affymetrix Rat Gene 1.0 ST Array 10795583 - -
          Affymetrix Rat Gene 1.0 ST Array 10795580 - -
          Affymetrix Rat Gene 1.0 ST Array 10795575 - -
          Affymetrix Rat Gene 1.0 ST Array 10795579 - -
          Affymetrix Rat Gene 1.0 ST Array 10795585 - -
          Affymetrix Rat Gene 1.0 ST Array 10795591 - -
          Affymetrix Rat Gene 1.0 ST Array 10795581 - -
          Affymetrix Rat Gene 1.0 ST Array 10795578 - -
          Affymetrix Rat Gene 1.0 ST Array 10795593 - -
          Affymetrix Rat Gene 1.1 ST Array 10795574 - -
          Rat Genome 230 2.0 Array 1391101_at - -
          Rat Genome 230 2.0 Array 1391101_PM_at - -
          Affymetrix HT Rat Focus Array 1391101_PM_at - -
          Affymetrix HT Rat Focus Array 1391101_at - -
          Affymetrix Rat Clariom S Assay PSR1700005307.rn.1 TC1700000784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700005306.rn.1 TC1700000784.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700005308.rn.1 TC1700000784.rn.1 RTA TC
          Rat Expression Set 230 1391101_PM_at - -
          Rat Expression Set 230 1391101_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795574 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20930885 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368216 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline