Search Thermo Fisher Scientific
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017279.1 | NP_058975.1 | 2 - 3 | 124 | 149 | |
GenBank mRNA | BC060576.1 | - | 2 - 3 | 143 | 6919255 | 149 |
FQ214776.1 | - | 2 - 3 | 139 | 149 | ||
FQ223872.1 | - | 139 | 149 | |||
J02897.1 | - | 2 - 3 | 124 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |