Search
Search
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017279.1 | NP_058975.1 | 5 - 6 | 469 | 93 | |
| GenBank mRNA | BC060576.1 | - | 5 - 6 | 488 | 6919255 | 93 |
| FQ214776.1 | - | 5 - 6 | 484 | 93 | ||
| FQ223872.1 | - | 484 | 93 | |||
| J02897.1 | - | 5 - 6 | 469 | 93 | ||
Set Membership: |
|