Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01542175_m1
          See other RIMS2 GE Assays ›
          Gene Symbol:
          Rims2
          Gene Name:
          regulating synaptic membrane exocytosis 2
          Gene Aliases:
          Nim2, Rim2
          Chromosome Location:
          Chr.7: 78092020 - 78594162 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01542175_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rims2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053945.2 NP_446397.1 25 - 26 4139 87
          NM_145881.2 NP_665888.1 2 - 3 461 87
          XM_006241591.2 XP_006241653.1 23 - 24 4207 87
          XM_006241592.2 XP_006241654.1 22 - 23 4141 87
          XM_006241593.2 XP_006241655.1 24 - 25 4107 87
          XM_006241594.2 XP_006241656.1 23 - 24 4065 87
          XM_006241598.2 XP_006241660.1 23 - 24 6911 87
          XM_008765437.1 XP_008763659.1 26 - 27 6409 87
          XM_008765438.1 XP_008763660.1 24 - 25 4258 87
          XM_008765439.1 XP_008763661.1 22 - 23 6103 87
          XM_008765440.1 XP_008763662.1 20 - 21 3688 87
          XM_008765441.1 XP_008763663.1 21 - 22 3595 87
          XM_008765442.1 XP_008763664.1 20 - 21 3547 87
          XM_008765443.1 XP_008763665.1 19 - 20 3480 87
          XM_008765445.1 XP_008763667.1 3 - 4 1389 87
          XM_008765446.1 XP_008763668.1 3 - 4 3322 87
          XM_008765447.1 XP_008763669.1 23 - 24 4081 87
          GenBank mRNA AF199322.1 - 25 - 26 4139 87
          AF199323.1 - 20 - 21 3530 87
          AF199324.1 - 20 - 21 3671 87
          AF199325.1 - 21 - 22 3578 87
          AF199326.1 - 20 - 21 3530 87
          AF199327.1 - 20 - 21 3530 87
          AF199328.1 - 19 - 20 3350 87
          AF199329.1 - 19 - 20 3464 87
          AF199330.1 - 24 - 25 4091 87
          AF199331.1 - 23 - 24 4049 87
          AF199332.1 - 20 - 21 3530 87
          AF199335.1 - 2 - 3 462 87
          AF548738.1 - 23 - 24 3562 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001018.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rims2

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          exocytosis
          positive regulation of gene expression
          calcium ion regulated exocytosis
          regulation of exocytosis
          cAMP-mediated signaling
          insulin secretion
          regulation of membrane potential
          calcium ion-regulated exocytosis of neurotransmitter
          positive regulation of inhibitory postsynaptic potential
          positive regulation of excitatory postsynaptic potential
          protein binding
          Rab GTPase binding
          protein domain specific binding
          ion channel binding
          metal ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0700007713.rn.1 TC0700001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007716.rn.1 TC0700001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007712.rn.1 TC0700001018.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline