Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01637108_m1
          See other SAG GE Assays ›
          Gene Symbol:
          Sag
          Gene Name:
          S-antigen; retina and pineal gland (arrestin)
          Gene Aliases:
          SAGMR, SANTI
          Chromosome Location:
          Chr.9: 94926838 - 94972162 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01637108_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sag
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013023.2 NP_037155.2 12 - 13 1149 85
          XM_008767196.1 XP_008765418.1 12 - 13 1412 85
          GenBank mRNA M60737.1 - 11 - 12 1040 85
          X15353.1 - 11 - 12 1040 85
          X51781.1 - 12 - 13 1149 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001086.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sag

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          visual perception
          biological_process
          opsin binding
          molecular_function
          spectrin binding
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054775 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840005 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150097 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136312 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054775 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054775 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018317 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018317 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054775 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P374851 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700301711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10448 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29760 - -
          Agilent Rat cDNA Microarray (G4105A) 700301711 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925112 - -
          Affymetrix Rat Gene 1.0 ST Array 10925116 - -
          Affymetrix Rat Gene 1.0 ST Array 10925115 - -
          Affymetrix Rat Gene 1.0 ST Array 10925125 - -
          Affymetrix Rat Gene 1.0 ST Array 10925114 - -
          Affymetrix Rat Gene 1.0 ST Array 10925122 - -
          Affymetrix Rat Gene 1.0 ST Array 10925119 - -
          Affymetrix Rat Gene 1.0 ST Array 10925126 - -
          Affymetrix Rat Gene 1.0 ST Array 10925118 - -
          Affymetrix Rat Gene 1.0 ST Array 10925123 - -
          Affymetrix Rat Gene 1.0 ST Array 10925120 - -
          Affymetrix Rat Gene 1.0 ST Array 10925124 - -
          Affymetrix Rat Gene 1.0 ST Array 10925117 - -
          Affymetrix Rat Gene 1.0 ST Array 10925121 - -
          Affymetrix Rat Gene 1.0 ST Array 10925113 - -
          Affymetrix Rat Gene 1.1 ST Array 10925111 - -
          Rat Genome 230 2.0 Array 1368112_PM_at - -
          Rat Genome 230 2.0 Array 1368112_at - -
          Affymetrix HT Rat Focus Array 1368112_at - -
          Affymetrix HT Rat Focus Array 1368112_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900009306.rn.1 TC0900001086.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900009299.rn.1 TC0900001086.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900009305.rn.1 TC0900001086.rn.1 RTA TC
          Rat Expression Set 230 1368112_at - -
          Rat Expression Set 230 1368112_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925111 - -
          Rat Genome U34 Set X51781cds_s_at - -
          Rat Genome U34 Set M60737_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21251635 - -
          ABI Rat Genome Survey Microarray 21386418 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361232 - -
          RatRef-12 Expression BeadChip ILMN_58249 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline