Search
Search
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017304.2 | NP_059000.1 | 5 - 6 | 681 | 63 | |
| XM_008764342.1 | XP_008762564.1 | 5 - 6 | 447 | 63 | ||
| XM_008764343.1 | XP_008762565.1 | 5 - 6 | 700 | 63 | ||
| XM_008764344.1 | XP_008762566.1 | 4 - 5 | 404 | 63 | ||
| XM_008764345.1 | XP_008762567.1 | 3 - 4 | 289 | 63 | ||
| GenBank mRNA | X76724.1 | - | 5 - 6 | 791 | 63 | |
Set Membership: |
|