Search
Search
| Gene Symbol: Atp5i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080481.1 | NP_536729.1 | 1 - 2 | 54 | 67 | |
| GenBank mRNA | BC058449.1 | - | 52 | 6890563 | 67 | |
| D13121.1 | - | 1 - 2 | 54 | 67 | ||
| FQ223740.1 | - | 1 - 2 | 44 | 67 | ||
Set Membership: |
|