Search
Search
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134465.2 | NP_604460.2 | 7 - 8 | 972 | 65 | |
| XM_008769915.1 | XP_008768137.1 | 7 - 8 | 930 | 65 | ||
| XM_008769916.1 | XP_008768138.1 | 7 - 8 | 955 | 65 | ||
| GenBank mRNA | AF404510.1 | - | 7 - 8 | 915 | 65 | |
| BC078740.1 | - | 7 - 8 | 972 | 7108137 | 65 | |
Set Membership: |
|