Search
Search
| Gene Symbol: Necap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199096.1 | NP_954527.1 | 6 - 7 | 683 | 62 | |
| XM_006239179.2 | XP_006239241.1 | 6 - 7 | 716 | 62 | ||
| GenBank mRNA | BC061826.1 | - | 6 - 7 | 683 | 5599028 | 62 |
Set Membership: |
|