Search
Search
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033235.1 | NP_150238.1 | 1 - 2 | 45 | 89 | |
| GenBank mRNA | AF093773.1 | - | 1 - 2 | 45 | 89 | |
| BC059124.1 | - | 1 - 2 | 86 | 6921518 | 89 | |
| FQ212702.1 | - | 1 - 2 | 89 | 89 | ||
| FQ215129.1 | - | 1 - 2 | 99 | 89 | ||
| FQ215603.1 | - | 1 - 2 | 84 | 89 | ||
| FQ217819.1 | - | 89 | 89 | |||
| FQ224891.1 | - | 1 - 2 | 87 | 89 | ||
| FQ233582.1 | - | 1 - 2 | 89 | 89 | ||
Set Membership: |
|