Search
Search
| Gene Symbol: Kcnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172041.2 | NP_742038.2 | 3 - 4 | 529 | 131 | |
| NM_172042.1 | NP_742039.1 | 3 - 4 | 517 | 131 | ||
| XM_006250433.2 | XP_006250495.1 | 3 - 4 | 740 | 131 | ||
| XM_006250434.2 | XP_006250496.1 | 3 - 4 | 1119 | 131 | ||
| GenBank mRNA | AF325671.1 | - | 3 - 4 | 517 | 131 | |
| AF385402.1 | - | 3 - 4 | 624 | 131 | ||
| AY555072.1 | - | 3 - 4 | 480 | 131 | ||
| AY555073.1 | - | 3 - 4 | 501 | 131 | ||
| AY695826.1 | - | 3 - 4 | 474 | 131 | ||
| AY727922.1 | - | 3 - 4 | 472 | 131 | ||
| DQ403851.1 | - | 3 - 4 | 529 | 131 | ||
Set Membership: |
|