Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01645767_m1
          See other FAM129A GE Assays ›
          Gene Symbol:
          Fam129a
          Gene Name:
          family with sequence similarity 129, member A
          Gene Aliases:
          Niban
          Chromosome Location:
          Chr.13: 68949665 - 69101600 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01645767_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam129a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022242.2 NP_071578.2 13 - 14 1696 143
          GenBank mRNA AB046718.2 - 13 - 14 1696 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000466.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam129a

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          response to endoplasmic reticulum stress
          positive regulation of translation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068228 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15226 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140343 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119742 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068222 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068228 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P633664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85163 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39665 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11053 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10764669 - -
          Affymetrix Rat Gene 1.0 ST Array 10764667 - -
          Affymetrix Rat Gene 1.0 ST Array 10764671 - -
          Affymetrix Rat Gene 1.0 ST Array 10764663 - -
          Affymetrix Rat Gene 1.0 ST Array 10764674 - -
          Affymetrix Rat Gene 1.0 ST Array 10764673 - -
          Affymetrix Rat Gene 1.0 ST Array 10764664 - -
          Affymetrix Rat Gene 1.0 ST Array 10764677 - -
          Affymetrix Rat Gene 1.0 ST Array 10764666 - -
          Affymetrix Rat Gene 1.0 ST Array 10764672 - -
          Affymetrix Rat Gene 1.0 ST Array 10764668 - -
          Affymetrix Rat Gene 1.0 ST Array 10764670 - -
          Affymetrix Rat Gene 1.0 ST Array 10764676 - -
          Affymetrix Rat Gene 1.0 ST Array 10764662 - -
          Affymetrix Rat Gene 1.0 ST Array 10764675 - -
          Affymetrix Rat Gene 1.0 ST Array 10764665 - -
          Affymetrix Rat Gene 1.1 ST Array 10764661 - -
          Rat Genome 230 2.0 Array 1387454_at - -
          Rat Genome 230 2.0 Array 1387454_PM_at - -
          Affymetrix HT Rat Focus Array 1387454_at - -
          Affymetrix HT Rat Focus Array 1387454_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300003270.rn.1 TC1300000466.rn.1 RTA TC
          Rat Expression Set 230 1387454_at - -
          Rat Expression Set 230 1387454_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10764661 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22005887 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366304 - -
          RatRef-12 Expression BeadChip ILMN_68421 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline