Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01646383_m1
          See other SV2C GE Assays ›
          Gene Symbol:
          Sv2c
          Gene Name:
          synaptic vesicle glycoprotein 2c
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 26502115 - 26699276 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01646383_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sv2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031593.1 NP_113781.1 12 - 13 2219 69
          GenBank mRNA AF060174.1 - 12 - 13 2219 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002677.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sv2c

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          transmembrane transport
          protein binding
          transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16768 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66099 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105824 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129936 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032946 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032946 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49003 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53660 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71462 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17501 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77574 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81446 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87800 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30695 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64916 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67824 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93138 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10820619 - -
          Affymetrix Rat Gene 1.0 ST Array 10820621 - -
          Affymetrix Rat Gene 1.0 ST Array 10820624 - -
          Affymetrix Rat Gene 1.0 ST Array 10820618 - -
          Affymetrix Rat Gene 1.0 ST Array 10820625 - -
          Affymetrix Rat Gene 1.0 ST Array 10820614 - -
          Affymetrix Rat Gene 1.0 ST Array 10820626 - -
          Affymetrix Rat Gene 1.0 ST Array 10820620 - -
          Affymetrix Rat Gene 1.0 ST Array 10820623 - -
          Affymetrix Rat Gene 1.0 ST Array 10820622 - -
          Affymetrix Rat Gene 1.0 ST Array 10820616 - -
          Affymetrix Rat Gene 1.0 ST Array 10820617 - -
          Affymetrix Rat Gene 1.0 ST Array 10820615 - -
          Affymetrix Rat Gene 1.1 ST Array 10820613 - -
          Rat Genome 230 2.0 Array 1387569_at - -
          Rat Genome 230 2.0 Array 1398221_PM_at - -
          Rat Genome 230 2.0 Array 1391071_at - -
          Rat Genome 230 2.0 Array 1398221_at - -
          Rat Genome 230 2.0 Array 1391071_PM_at - -
          Rat Genome 230 2.0 Array 1387569_PM_at - -
          Affymetrix HT Rat Focus Array 1387569_PM_at - -
          Affymetrix HT Rat Focus Array 1387569_at - -
          Affymetrix Rat Clariom S Assay PSR0200018704.rn.1 TC0200002677.rn.1 RTA TC
          Rat Expression Set 230 1387569_at - -
          Rat Expression Set 230 1387569_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10820613 - -
          Rat Genome U34 Set AF060174_at - -
          Rat Genome U34 Set AF060174_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21961299 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355784 - -
          RatRef-12 Expression BeadChip ILMN_50955 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline