Search
Search
| Gene Symbol: Stx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031125.1 | NP_112387.1 | 8 - 9 | 734 | 98 | |
| XM_006230345.2 | XP_006230407.1 | 8 - 9 | 1241 | 98 | ||
| XM_006230346.2 | XP_006230408.1 | 8 - 9 | 1266 | 98 | ||
| GenBank mRNA | L20821.1 | - | 8 - 9 | 734 | 98 | |
Set Membership: |
|