Search
Search
| Gene Symbol: Gab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053417.1 | NP_445869.1 | 5 - 6 | 1272 | 110 | |
| XM_008759806.1 | XP_008758028.1 | 4 - 5 | 2904 | 110 | ||
| GenBank mRNA | AF230367.2 | - | 5 - 6 | 1272 | 110 | |
Set Membership: |
|