Search
Search
| Gene Symbol: Cry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198750.2 | NP_942045.2 | 4 - 5 | 1132 | 85 | |
| XM_008765225.1 | XP_008763447.1 | 4 - 5 | 1156 | 85 | ||
| GenBank mRNA | AF545855.1 | - | 4 - 5 | 696 | 85 | |
| BC107677.1 | - | 4 - 5 | 1132 | 7939194 | 85 | |
Set Membership: |
|