Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01746678_m1
          See other CDV3 GE Assays ›
          Gene Symbol:
          Cdv3
          Gene Name:
          carnitine deficiency-associated gene expressed in ventricle 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 111837235 - 111850393 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01746678_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdv3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014097.1 NP_001014119.1 2 - 3 795 96
          XM_006243659.2 XP_006243721.1 2 - 3 701 96
          XM_006243660.2 XP_006243722.1 2 - 3 699 96
          XM_008766529.1 XP_008764751.1 2 - 3 630 96
          GenBank mRNA BC083654.1 - 2 - 3 795 7135192 96

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_008766529.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdv3

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1014379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136612 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1014379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1014379 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1014379 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1014383 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1014379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919685 - -
          Affymetrix Rat Gene 1.0 ST Array 10919682 - -
          Affymetrix Rat Gene 1.0 ST Array 10919686 - -
          Affymetrix Rat Gene 1.0 ST Array 10919684 - -
          Affymetrix Rat Gene 1.0 ST Array 10919683 - -
          Affymetrix Rat Gene 1.0 ST Array 10724639 - -
          Affymetrix Rat Gene 1.1 ST Array 10724637 - -
          Affymetrix Rat Gene 1.1 ST Array 10919681 - -
          Rat Genome 230 2.0 Array 1383028_PM_at - -
          Rat Genome 230 2.0 Array 1383028_at - -
          Affymetrix HT Rat Focus Array 1383028_PM_at - -
          Affymetrix HT Rat Focus Array 1383028_at - -
          Affymetrix Rat Clariom S Assay PSR0800026085.rn.1 TC0800003275.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800026087.rn.1 TC0800003275.rn.1 RTA TC
          Rat Expression Set 230 1383028_PM_at - -
          Rat Expression Set 230 1383028_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919681 - -
          Rat Genome U34 Set rc_AI639455_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21849579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline