Search
Search
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025689.1 | NP_001020860.1 | 5 - 6 | 522 | 76 | |
| GenBank mRNA | BC097427.1 | - | 5 - 6 | 522 | 76 | |
| BC158645.1 | - | 5 - 6 | 470 | 8363877 | 76 | |
| FQ228827.1 | - | 5 - 6 | 498 | 76 | ||
Set Membership: |
|