Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01747638_m1
          See other PRR5L GE Assays ›
          Gene Symbol:
          Prr5l
          Gene Name:
          proline rich 5 like
          Gene Aliases:
          Protor-2, Protor2, RGD1309969
          Chromosome Location:
          Chr.3: 91291347 - 91370943 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01747638_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr5l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080150.1 NP_001073619.1 2 - 3 523 71
          XM_006234647.2 XP_006234709.1 2 - 3 1489 71
          XM_006234648.2 XP_006234710.1 2 - 3 790 71
          GenBank mRNA BC129104.1 - 2 - 3 523 7100178 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003749.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          biological_process
          negative regulation of signal transduction
          regulation of fibroblast migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          TORC2 signaling
          molecular_function
          ubiquitin protein ligase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P124838 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P634470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67385 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122626 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P634470 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P634470 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124838 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P634470 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P124838 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P173299 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P634470 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75542 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87509 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62167 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10847744 - -
          Affymetrix Rat Gene 1.0 ST Array 10847743 - -
          Affymetrix Rat Gene 1.0 ST Array 10847736 - -
          Affymetrix Rat Gene 1.0 ST Array 10847738 - -
          Affymetrix Rat Gene 1.0 ST Array 10847741 - -
          Affymetrix Rat Gene 1.0 ST Array 10847740 - -
          Affymetrix Rat Gene 1.0 ST Array 10847742 - -
          Affymetrix Rat Gene 1.0 ST Array 10847737 - -
          Affymetrix Rat Gene 1.0 ST Array 10847739 - -
          Affymetrix Rat Gene 1.1 ST Array 10847735 - -
          Rat Genome 230 2.0 Array 1376775_PM_at - -
          Rat Genome 230 2.0 Array 1376775_at - -
          Affymetrix HT Rat Focus Array 1376775_at - -
          Affymetrix HT Rat Focus Array 1376775_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300028497.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028503.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028499.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028507.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028511.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028498.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028500.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028504.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028506.rn.1 TC0300003749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028510.rn.1 TC0300003749.rn.1 RTA TC
          Rat Expression Set 230 1376775_PM_at - -
          Rat Expression Set 230 1376775_at - -
          Affymetrix Rat Gene 1.0 ST Array 10847735 - -
          Rat Genome U34 Set rc_AI059078_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651021 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline