Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01748352_m1
          See other SLC39A8 GE Assays ›
          Gene Symbol:
          Slc39a8
          Gene Name:
          solute carrier family 39 (zinc transporter), member 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 241029188 - 241092584 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01748352_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc39a8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011952.1 NP_001011952.1 7 - 8 1492 67
          XM_006233356.2 XP_006233418.1 8 - 9 1531 67
          XM_006233357.2 XP_006233419.1 8 - 9 1751 67
          GenBank mRNA BC089844.1 - 7 - 8 1492 7368889 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002265.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc39a8

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          zinc II ion transport
          cadmium ion transmembrane transport
          metal ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1052361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87664 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54841 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1052361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1052361 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1052361 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1052361 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P278975 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66712 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78116 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6385 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819273 - -
          Affymetrix Rat Gene 1.0 ST Array 10819280 - -
          Affymetrix Rat Gene 1.0 ST Array 10819276 - -
          Affymetrix Rat Gene 1.0 ST Array 10819279 - -
          Affymetrix Rat Gene 1.0 ST Array 10819271 - -
          Affymetrix Rat Gene 1.0 ST Array 10819272 - -
          Affymetrix Rat Gene 1.0 ST Array 10819274 - -
          Affymetrix Rat Gene 1.0 ST Array 10819278 - -
          Affymetrix Rat Gene 1.0 ST Array 10819270 - -
          Affymetrix Rat Gene 1.1 ST Array 10819269 - -
          Rat Genome 230 2.0 Array 1376709_at - -
          Rat Genome 230 2.0 Array 1376709_PM_at - -
          Affymetrix HT Rat Focus Array 1376709_PM_at - -
          Affymetrix HT Rat Focus Array 1376709_at - -
          Affymetrix Rat Clariom S Assay PSR0200015717.rn.1 TC0200002265.rn.1 RTA TC
          Rat Expression Set 230 1376709_PM_at - -
          Rat Expression Set 230 1376709_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819269 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22312843 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376054 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376355 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline