Search
Search
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007699.1 | NP_001007700.1 | 4 - 5 | 863 | 86 | |
| XM_006233116.2 | XP_006233178.1 | 4 - 5 | 791 | 86 | ||
| XM_006233118.2 | XP_006233180.1 | 4 - 5 | 1628 | 86 | ||
| GenBank mRNA | BC079471.1 | - | 4 - 5 | 863 | 7122765 | 86 |
Set Membership: |
|