Search
Search
| Gene Symbol: Cd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024335.1 | NP_001019506.1 | 4 - 5 | 581 | 62 | |
| XM_006237380.2 | XP_006237442.1 | 4 - 5 | 2091 | 62 | ||
| XM_006237382.2 | XP_006237444.1 | 4 - 5 | 2206 | 62 | ||
| XM_008763293.1 | XP_008761515.1 | 5 - 6 | 1886 | 62 | ||
| GenBank mRNA | BC095844.1 | - | 4 - 5 | 581 | 7389613 | 62 |
Set Membership: |
|