Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107903.1 | NP_001101373.1 | 3 - 4 | 564 | 66 | |
XM_006237827.2 | XP_006237889.1 | 3 - 4 | 593 | 66 | ||
XM_006237832.2 | XP_006237894.1 | 3 - 4 | 594 | 66 | ||
XM_008763539.1 | XP_008761761.1 | 3 - 4 | 599 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |