Search
Search

Slco5a1| Gene Symbol: Slco5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107898.1 | NP_001101368.1 | 7 - 8 | 2650 | 61 | |
| XM_006237743.2 | XP_006237805.1 | 8 - 9 | 2765 | 61 | ||
| XM_008763495.1 | XP_008761717.1 | 8 - 9 | 3071 | 61 | ||
Set Membership: |
|