Search
Search
| Gene Symbol: Nfatc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107805.1 | NP_001101275.1 | 4 - 5 | 1623 | 68 | |
| XM_006235653.1 | XP_006235715.1 | 4 - 5 | 1617 | 68 | ||
| XM_008762507.1 | XP_008760729.1 | 4 - 5 | 1617 | 68 | ||
Set Membership: |
|