Search
Search
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039341.1 | NP_001034430.1 | 8 - 9 | 1488 | 168 | |
| XM_006247070.2 | XP_006247132.1 | 8 - 9 | 1683 | 128 | ||
| GenBank mRNA | BC110049.1 | - | 8 - 9 | 1488 | 7316890 | 168 |
Set Membership: |
|