Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prss33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109027.1 | NP_001102497.1 | 1 - 2 | 68 | 78 | |
XM_006245874.2 | XP_006245936.1 | 2 - 3 | 358 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |