Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751474_m1
          See other IGTP GE Assays ›
          Gene Symbol:
          Igtp
          Gene Name:
          interferon gamma induced GTPase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 43631086 - 43639044 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751474_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igtp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008765.1 NP_001008765.1 2 - 3 399 85
          XM_006246382.2 XP_006246444.1 2 - 3 418 85
          GenBank mRNA BC129105.1 - 2 - 3 399 85
          BC166569.1 - 2 - 3 400 7319275 85

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC129105.1, BC166569.1, NM_001008765.1, XM_006246382.2 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058492 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092170 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P179956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359022 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66798 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058492 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092170 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058492 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092170 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058492 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092170 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13862 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733801 - -
          Affymetrix Rat Gene 1.0 ST Array 10733798 - -
          Affymetrix Rat Gene 1.0 ST Array 10733800 - -
          Affymetrix Rat Gene 1.0 ST Array 10733799 - -
          Affymetrix Rat Gene 1.0 ST Array 10733802 - -
          Affymetrix Rat Gene 1.1 ST Array 10733790 - -
          Rat Genome 230 2.0 Array 1375796_PM_at - -
          Rat Genome 230 2.0 Array 1397304_PM_at - -
          Rat Genome 230 2.0 Array 1396163_at - -
          Rat Genome 230 2.0 Array 1397304_at - -
          Rat Genome 230 2.0 Array 1396163_PM_at - -
          Rat Genome 230 2.0 Array 1375796_at - -
          Affymetrix HT Rat Focus Array 1396163_at - -
          Affymetrix HT Rat Focus Array 1397304_at - -
          Affymetrix HT Rat Focus Array 1397304_PM_at - -
          Affymetrix HT Rat Focus Array 1396163_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000005674.rn.1 TC1000000666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000005667.rn.1 TC1000000666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000005673.rn.1 TC1000000666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000005672.rn.1 TC1000000666.rn.1 RTA TC
          Rat Expression Set 230 1375796_PM_at - -
          Rat Expression Set 230 1375796_at - -
          Rat Expression Set 230 1396163_at - -
          Rat Expression Set 230 1396163_PM_at - -
          Rat Expression Set 230 1397304_PM_at - -
          Rat Expression Set 230 1397304_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733790 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21377958 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349575 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline