Search
Search
| Gene Symbol: Sp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107045.1 | NP_001100515.1 | 5 - 6 | 1630 | 77 | |
| XM_006247303.2 | XP_006247365.1 | 4 - 5 | 1392 | 77 | ||
| GenBank mRNA | BC168669.1 | - | 4 - 5 | 1398 | 7934743 | 77 |
Set Membership: |
|