Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751772_g1
          See other LPO GE Assays ›
          Gene Symbol:
          Lpo
          Gene Name:
          lactoperoxidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 75100385 - 75120320 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751772_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lpo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105829.1 NP_001099299.1 12 - 13 2285 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003494.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          response to oxidative stress
          thiocyanate metabolic process
          defense response to bacterium
          oxidation-reduction process
          peroxidase activity
          heme binding
          thiocyanate peroxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18000 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70063 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155795 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155795 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155795 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501937 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746064 - -
          Affymetrix Rat Gene 1.0 ST Array 10746062 - -
          Affymetrix Rat Gene 1.0 ST Array 10746063 - -
          Affymetrix Rat Gene 1.0 ST Array 10746061 - -
          Affymetrix Rat Gene 1.0 ST Array 10746059 - -
          Affymetrix Rat Gene 1.0 ST Array 10746054 - -
          Affymetrix Rat Gene 1.0 ST Array 10746057 - -
          Affymetrix Rat Gene 1.0 ST Array 10746058 - -
          Affymetrix Rat Gene 1.0 ST Array 10746055 - -
          Affymetrix Rat Gene 1.0 ST Array 10746065 - -
          Affymetrix Rat Gene 1.0 ST Array 10746060 - -
          Affymetrix Rat Gene 1.0 ST Array 10746056 - -
          Affymetrix Rat Gene 1.1 ST Array 10746053 - -
          Rat Genome 230 2.0 Array 1377429_at - -
          Rat Genome 230 2.0 Array 1377429_PM_at - -
          Affymetrix HT Rat Focus Array 1377429_PM_at - -
          Affymetrix HT Rat Focus Array 1377429_at - -
          Affymetrix Rat Clariom S Assay PSR1000031313.rn.1 TC1000003494.rn.1 RTA TC
          Rat Expression Set 230 1377429_at - -
          Rat Expression Set 230 1377429_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10746053 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21396384 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362386 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline