Search
Search
| Gene Symbol: Myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106017.1 | NP_001099487.1 | 1 - 2 | 124 | 62 | |
| XM_006251216.2 | XP_006251278.1 | 2 - 3 | 224 | 62 | ||
| GenBank mRNA | BC161995.1 | - | 124 | 9034230 | 62 | |
Set Membership: |
|