Search
Search

Igf2bp2| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270598.1 | NP_001257527.1 | 10 - 11 | 1264 | 124 | |
| NM_001270599.1 | NP_001257528.1 | 10 - 11 | 1264 | 124 | ||
| GenBank mRNA | FQ222567.1 | - | 10 - 11 | 1032 | 124 | |
Set Membership: |
|