Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753476_g1
          See other GGACT GE Assays ›
          Gene Symbol:
          Ggact
          Gene Name:
          gamma-glutamylamine cyclotransferase
          Gene Aliases:
          A2ld1, RGD1304748
          Chromosome Location:
          Chr.15: 109307683 - 109336029 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753476_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ggact
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025634.1 NP_001020805.1 3 - 4 199 88
          XM_006252512.2 XP_006252574.1 3 - 4 1532 88
          XM_006252513.2 XP_006252575.1 3 - 4 402 88
          XM_006252514.2 XP_006252576.1 3 - 4 594 88
          XM_006252515.2 XP_006252577.1 3 - 4 652 88
          XM_006252516.2 XP_006252578.1 2 - 3 1448 88
          XM_006252520.2 XP_006252582.1 3 - 4 505 88
          XM_006252521.2 XP_006252583.1 2 - 3 318 88
          XM_006252522.2 XP_006252584.1 2 - 3 510 88
          XM_008770970.1 XP_008769192.1 3 - 4 1538 88
          XM_008770971.1 XP_008769193.1 2 - 3 1843 88
          XM_008770972.1 XP_008769194.1 2 - 3 942 88
          GenBank mRNA BC098699.1 - 3 - 4 199 7386922 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the BC098699.1,NM_001025634.1,XM_006252515.2,XM_006252516.2,XM_006252520.2,XM_006252521.2,XM_006252522.2,XM_008770970.1,XM_008770971.1,XM_008770972.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ggact

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular modified amino acid catabolic process
          gamma-glutamylcyclotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150164 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101912 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005832 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005832 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005832 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P197807 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536665 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51476 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786038 - -
          Affymetrix Rat Gene 1.0 ST Array 10786041 - -
          Affymetrix Rat Gene 1.0 ST Array 10786037 - -
          Affymetrix Rat Gene 1.0 ST Array 10786039 - -
          Affymetrix Rat Gene 1.0 ST Array 10786040 - -
          Affymetrix Rat Gene 1.1 ST Array 10786036 - -
          Rat Genome 230 2.0 Array 1382945_at - -
          Rat Genome 230 2.0 Array 1392725_at - -
          Rat Genome 230 2.0 Array 1382945_PM_at - -
          Rat Genome 230 2.0 Array 1392725_PM_at - -
          Affymetrix HT Rat Focus Array 1392725_at - -
          Affymetrix HT Rat Focus Array 1392725_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500016386.rn.1 TC1500002448.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016391.rn.1 TC1500002448.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016387.rn.1 TC1500002448.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016385.rn.1 TC1500002448.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016388.rn.1 TC1500002448.rn.1 RTA TC
          Rat Expression Set 230 1392725_at - -
          Rat Expression Set 230 1382945_at - -
          Rat Expression Set 230 1382945_PM_at - -
          Rat Expression Set 230 1392725_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786036 - -
          Rat Genome U34 Set rc_AA943039_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21872972 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357910 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline