Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753527_m1
          See other ITGBL1 GE Assays ›
          Gene Symbol:
          Itgbl1
          Gene Name:
          integrin, beta-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 110114148 - 110378970 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753527_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgbl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017505.1 NP_001017505.1 10 - 11 1480 89
          GenBank mRNA BC087076.1 - 10 - 11 1480 7134947 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001218.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgbl1

          Panther Classification:

          Molecular Function -

          integrin

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P106573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151809 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61981 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48080 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106574 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106574 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P106574 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66857 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2387 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38641 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76128 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19035 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21061 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55998 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47529 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77163 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782196 - -
          Affymetrix Rat Gene 1.0 ST Array 10782193 - -
          Affymetrix Rat Gene 1.0 ST Array 10782194 - -
          Affymetrix Rat Gene 1.0 ST Array 10782192 - -
          Affymetrix Rat Gene 1.0 ST Array 10782195 - -
          Affymetrix Rat Gene 1.0 ST Array 10782188 - -
          Affymetrix Rat Gene 1.0 ST Array 10782189 - -
          Affymetrix Rat Gene 1.0 ST Array 10782197 - -
          Affymetrix Rat Gene 1.0 ST Array 10782198 - -
          Affymetrix Rat Gene 1.0 ST Array 10782191 - -
          Affymetrix Rat Gene 1.0 ST Array 10782190 - -
          Affymetrix Rat Gene 1.1 ST Array 10782187 - -
          Rat Genome 230 2.0 Array 1383708_PM_at - -
          Rat Genome 230 2.0 Array 1383708_at - -
          Affymetrix HT Rat Focus Array 1383708_PM_at - -
          Affymetrix HT Rat Focus Array 1383708_at - -
          Affymetrix Rat Clariom S Assay PSR1500007633.rn.1 TC1500001218.rn.1 RTA TC
          Rat Expression Set 230 1383708_PM_at - -
          Rat Expression Set 230 1383708_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782187 - -
          Rat Genome U34 Set rc_AI235334_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21618568 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650212 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline