Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756153_g1
          See other DYNLT3 GE Assays ›
          Gene Symbol:
          Dynlt3
          Gene Name:
          dynein light chain Tctex-type 3
          Gene Aliases:
          Tcte1l
          Chromosome Location:
          Chr.X: 14633342 - 14642356 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756153_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dynlt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013228.1 NP_001013246.1 4 - 5 305 80
          GenBank mRNA BC083797.1 - 4 - 5 305 7108507 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001471.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dynlt3

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of mitotic cell cycle
          protein binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66163 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051034 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051034 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051034 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16703 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83447 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10932369 - -
          Affymetrix Rat Gene 1.0 ST Array 10932370 - -
          Affymetrix Rat Gene 1.0 ST Array 10932367 - -
          Affymetrix Rat Gene 1.0 ST Array 10932368 - -
          Affymetrix Rat Gene 1.0 ST Array 10932371 - -
          Affymetrix Rat Gene 1.1 ST Array 10932366 - -
          Rat Genome 230 2.0 Array 1388903_PM_at - -
          Rat Genome 230 2.0 Array 1388903_at - -
          Affymetrix HT Rat Focus Array 1388903_at - -
          Affymetrix HT Rat Focus Array 1388903_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00009329.rn.1 TC0X00001471.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00009330.rn.1 TC0X00001471.rn.1 RTA TC
          Rat Expression Set 230 1388903_at - -
          Rat Expression Set 230 1388903_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10932366 - -
          Rat Genome U34 Set rc_AI179335_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20858215 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651019 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline