Search
Search

Cyp39a1| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106893.1 | NP_001100363.1 | 6 - 7 | 1150 | 76 | |
| XM_006244604.2 | XP_006244666.1 | 6 - 7 | 1150 | 76 | ||
| XM_006244605.1 | XP_006244667.1 | 6 - 7 | 1150 | 76 | ||
| XM_008766854.1 | XP_008765076.1 | 6 - 7 | 1033 | 76 | ||
| XM_008766855.1 | XP_008765077.1 | 6 - 7 | 1031 | 76 | ||
Set Membership: |
|