Search
Search
| Gene Symbol: Haus7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277272.1 | NP_001264201.1 | 315 | 82 | ||
| XM_006229555.2 | XP_006229617.1 | 804 | 82 | |||
| GenBank mRNA | BC103640.1 | - | 298 | 7134953 | 82 | |
| BC158688.1 | - | 247 | 8375819 | 82 | ||
Set Membership: |
|