Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757475_m1
          See other FBXW17 GE Assays ›
          Gene Symbol:
          Fbxw17
          Gene Name:
          F-box and WD-40 domain protein 17
          Gene Aliases:
          RGD1566133
          Chromosome Location:
          Chr.17: 15076773 - 15094620 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01757475_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxw17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001082409.1 NP_001075878.1 9 - 10 1310 88
          XM_008771540.1 XP_008769762.1 8 - 9 1430 88
          XM_008771541.1 XP_008769763.1 8 - 9 1379 88
          GenBank mRNA BC167744.1 - 9 - 10 1293 7099140 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001429.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1003716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904450 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1003716 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1003716 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1003716 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1003716 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1003715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794277 - -
          Affymetrix Rat Gene 1.0 ST Array 10794283 - -
          Affymetrix Rat Gene 1.0 ST Array 10794282 - -
          Affymetrix Rat Gene 1.0 ST Array 10794280 - -
          Affymetrix Rat Gene 1.0 ST Array 10794285 - -
          Affymetrix Rat Gene 1.0 ST Array 10794286 - -
          Affymetrix Rat Gene 1.0 ST Array 10794281 - -
          Affymetrix Rat Gene 1.0 ST Array 10794279 - -
          Affymetrix Rat Gene 1.0 ST Array 10794284 - -
          Affymetrix Rat Gene 1.0 ST Array 10794278 - -
          Affymetrix Rat Gene 1.1 ST Array 10794276 - -
          Rat Genome 230 2.0 Array 1384457_at - -
          Rat Genome 230 2.0 Array 1396671_at - -
          Rat Genome 230 2.0 Array 1396671_PM_at - -
          Rat Genome 230 2.0 Array 1384457_PM_at - -
          Affymetrix HT Rat Focus Array 1384457_PM_at - -
          Affymetrix HT Rat Focus Array 1384457_at - -
          Affymetrix Rat Clariom S Assay PSR1700009950.rn.1 TC1700001429.rn.1 RTA TC
          Rat Expression Set 230 1384457_PM_at - -
          Rat Expression Set 230 1384457_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794276 - -
          Rat Genome U34 Set rc_AI030755_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22240612 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374294 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358797 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline