Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01758621_g1
See other NOTCH1 GE Assays ›
Gene Symbol:
Notch1
Gene Name:
notch 1
Gene Aliases:
NOTCH, TAN1
Chromosome Location:
Chr.3: 3905562 - 3951015 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01758621_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Notch1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001105721.1
NP_001099191.1
20 - 21
3549
75
GenBank mRNA
X57405.1
-
20 - 21
3563
75
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300002730.rn.1
Set Membership:
Amplicon length between 71 and 85
Detect Genomic DNA
Rattus norvegicus
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
cell fate specification
epithelial to mesenchymal transition
liver development
heart looping
sprouting angiogenesis
positive regulation of neuroblast proliferation
inflammatory response to antigenic stimulus
endocardium development
endocardium morphogenesis
atrioventricular node development
coronary vein morphogenesis
aortic valve morphogenesis
atrioventricular valve morphogenesis
pulmonary valve morphogenesis
mitral valve formation
epithelial to mesenchymal transition involved in endocardial cushion formation
endocardial cushion morphogenesis
cardiac chamber formation
cardiac ventricle morphogenesis
cardiac atrium morphogenesis
cardiac right atrium morphogenesis
cardiac left ventricle morphogenesis
cardiac right ventricle formation
ventricular trabecula myocardium morphogenesis
growth involved in heart morphogenesis
regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
regulation of cardioblast proliferation
Notch signaling pathway involved in regulation of secondary heart field cardioblast proliferation
cell migration involved in endocardial cushion formation
pericardium morphogenesis
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
humoral immune response
Notch signaling pathway
positive regulation of transcription of Notch receptor target
spermatogenesis
determination of left/right symmetry
compartment pattern specification
axonogenesis
brain development
foregut morphogenesis
endoderm development
heart development
positive regulation of cell proliferation
negative regulation of cell proliferation
epidermis development
auditory receptor cell fate commitment
glial cell differentiation
regulation of gene expression
positive regulation of epithelial to mesenchymal transition
negative regulation of cell-substrate adhesion
negative regulation of myotube differentiation
mesenchymal cell development
regulation of somitogenesis
cell differentiation in spinal cord
neural tube development
cell differentiation
neuron differentiation
keratinocyte differentiation
negative regulation of ossification
lung development
embryonic limb morphogenesis
regulation of cell migration
positive regulation of cell migration
positive regulation of BMP signaling pathway
negative regulation of BMP signaling pathway
forebrain development
hair follicle morphogenesis
organ regeneration
response to corticosteroid
response to muramyl dipeptide
response to lipopolysaccharide
embryonic hindlimb morphogenesis
tube formation
skeletal muscle cell differentiation
cellular response to vascular endothelial growth factor stimulus
regulation of cell proliferation
tissue regeneration
positive regulation of apoptotic process
negative regulation of catalytic activity
cell fate commitment
negative regulation of cell differentiation
positive regulation of endothelial cell differentiation
positive regulation of keratinocyte differentiation
negative regulation of myoblast differentiation
negative regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of glial cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of calcium ion-dependent exocytosis
positive regulation of JAK-STAT cascade
negative regulation of photoreceptor cell differentiation
somatic stem cell division
neuron fate commitment
astrocyte differentiation
oligodendrocyte differentiation
positive regulation of astrocyte differentiation
negative regulation of oligodendrocyte differentiation
branching morphogenesis of an epithelial tube
regulation of epithelial cell proliferation
positive regulation of epithelial cell proliferation
regulation of neurogenesis
negative regulation of neurogenesis
cardiac muscle tissue morphogenesis
cardiac muscle cell proliferation
positive regulation of cardiac muscle cell proliferation
negative regulation of glial cell proliferation
cilium morphogenesis
cardiac epithelial to mesenchymal transition
cardiac septum morphogenesis
ventricular septum morphogenesis
secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development
negative regulation of cell death
prostate gland epithelium morphogenesis
regulation of epithelial cell proliferation involved in prostate gland development
arterial endothelial cell differentiation
venous endothelial cell differentiation
cardiac vascular smooth muscle cell development
endocardial cell differentiation
vasculogenesis involved in coronary vascular morphogenesis
coronary artery morphogenesis
Notch signaling involved in heart development
heart trabecula morphogenesis
positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
left/right axis specification
cellular response to follicle-stimulating hormone stimulus
distal tubule development
collecting duct development
glomerular mesangial cell development
interleukin-4 secretion
negative regulation of cell migration involved in sprouting angiogenesis
negative regulation of canonical Wnt signaling pathway
neuronal stem cell population maintenance
regulation of extracellular matrix assembly
apoptotic process involved in embryonic digit morphogenesis
negative regulation of stem cell differentiation
negative regulation of anoikis
negative regulation of pro-B cell differentiation
negative regulation of endothelial cell chemotaxis
core promoter binding
transcriptional activator activity, RNA polymerase II transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
enzyme inhibitor activity
receptor activity
calcium ion binding
enzyme binding
chromatin DNA binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0300020658.rn.1
TC0300002730.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline