Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01759587_m1
          See other CYP2E1 GE Assays ›
          Gene Symbol:
          Cyp2e1
          Gene Name:
          cytochrome P450, family 2, subfamily e, polypeptide 1
          Gene Aliases:
          Cyp2e
          Chromosome Location:
          Chr.1: 213511538 - 213522196 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01759587_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2e1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031543.1 NP_113731.1 8 - 9 1309 126
          XM_006230520.1 XP_006230582.1 9 - 10 1492 126
          GenBank mRNA AF056333.1 - 3 - 4 391 126
          AF061442.1 - 8 - 9 1297 126
          BC081774.1 - 8 - 9 1311 7110096 126
          FQ209397.1 - 8 - 9 1328 126
          FQ209597.1 - 3 - 4 428 126
          FQ210662.1 - 3 - 4 428 126
          FQ210815.1 - 8 - 9 1330 126
          FQ211086.1 - 8 - 9 1330 126
          FQ212595.1 - 5 - 6 675 126
          FQ218532.1 - 8 - 9 1309 126
          FQ219736.1 - 8 - 9 1328 126
          J02627.1 - 8 - 9 1309 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002862.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2e1

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          triglyceride metabolic process
          xenobiotic metabolic process
          steroid metabolic process
          response to ozone
          response to organonitrogen compound
          monoterpenoid metabolic process
          drug metabolic process
          epoxygenase P450 pathway
          response to drug
          exogenous drug catabolic process
          response to ethanol
          heterocycle metabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          FMN binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          enzyme binding
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P409232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92515 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409232 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P409232 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39312 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712096 - -
          Affymetrix Rat Gene 1.0 ST Array 10712093 - -
          Affymetrix Rat Gene 1.0 ST Array 10712100 - -
          Affymetrix Rat Gene 1.0 ST Array 10712095 - -
          Affymetrix Rat Gene 1.0 ST Array 10712091 - -
          Affymetrix Rat Gene 1.0 ST Array 10712094 - -
          Affymetrix Rat Gene 1.0 ST Array 10712099 - -
          Affymetrix Rat Gene 1.0 ST Array 10712097 - -
          Affymetrix Rat Gene 1.0 ST Array 10712098 - -
          Affymetrix Rat Gene 1.0 ST Array 10712092 - -
          Affymetrix Rat Gene 1.1 ST Array 10712090 - -
          Rat Genome 230 2.0 Array 1367871_PM_at - -
          Rat Genome 230 2.0 Array 1367871_at - -
          Affymetrix HT Rat Focus Array 1367871_at - -
          Affymetrix HT Rat Focus Array 1367871_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100022524.rn.1 TC0100002862.rn.1 RTA TC
          Rat Expression Set 230 1367871_at - -
          Rat Expression Set 230 1367871_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10712090 - -
          Rat Genome U34 Set S48325_s_at - -
          Rat Genome U34 Set M20131cds_s_at - -
          Rat Genome U34 Set AF056333_s_at - -
          Rat Genome U34 Set rc_AA819450_i_at - -
          Rat Genome U34 Set rc_AA819450_f_at - -
          Rat Genome U34 Set M20131cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21754868 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358592 - -
          RatRef-12 Expression BeadChip ILMN_51303 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline