Search
Search
| Gene Symbol: Ric8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100520.1 | NP_001093990.1 | 5 - 6 | 1197 | 65 | |
| GenBank mRNA | AY177754.1 | - | 5 - 6 | 957 | 65 | |
| BC160852.1 | - | 5 - 6 | 1197 | 8364542 | 65 | |
| FQ223245.1 | - | 892 | 65 | |||
Set Membership: |
|