Search
Search
| Gene Symbol: Dnaaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014154.1 | NP_001014176.1 | 7 - 8 | 1063 | 105 | |
| GenBank mRNA | BC079038.1 | - | 7 - 8 | 1063 | 7114363 | 105 |
Set Membership: |
|