Search
Search
| Gene Symbol: Irf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008722.1 | NP_001008722.1 | 2 - 3 | 361 | 73 | |
| XM_006255737.2 | XP_006255799.1 | 3 - 4 | 409 | 73 | ||
| GenBank mRNA | AB120371.1 | - | 2 - 3 | 361 | 73 | |
Set Membership: |
|