Search
Search
| Gene Symbol: Zfp90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025765.1 | NP_001020936.1 | 2 - 3 | 174 | 87 | |
| XM_006255567.2 | XP_006255629.1 | 2 - 3 | 325 | 87 | ||
| GenBank mRNA | BC097467.1 | - | 2 - 3 | 174 | 7457672 | 87 |
Set Membership: |
|