Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01762563_m1
          See other MMAA GE Assays ›
          Gene Symbol:
          Mmaa
          Gene Name:
          methylmalonic aciduria (cobalamin deficiency) cblA type
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 32294087 - 32325927 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01762563_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmaa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106174.1 NP_001099644.1 7 - 8 1395 70
          XM_006255398.2 XP_006255460.1 6 - 7 1077 70
          XM_006255399.2 XP_006255461.1 6 - 7 3216 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000426.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          isomerase mutase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P806847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P806847 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P806847 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P806847 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1018181 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74702 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46639 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806998 - -
          Affymetrix Rat Gene 1.0 ST Array 10806999 - -
          Affymetrix Rat Gene 1.0 ST Array 10806994 - -
          Affymetrix Rat Gene 1.0 ST Array 10806995 - -
          Affymetrix Rat Gene 1.0 ST Array 10806997 - -
          Affymetrix Rat Gene 1.0 ST Array 10806996 - -
          Affymetrix Rat Gene 1.1 ST Array 10806993 - -
          Rat Genome 230 2.0 Array 1377302_PM_a_at - -
          Rat Genome 230 2.0 Array 1377302_a_at - -
          Affymetrix HT Rat Focus Array 1377302_PM_a_at - -
          Affymetrix HT Rat Focus Array 1377302_a_at - -
          Affymetrix Rat Clariom S Assay PSR1900003378.rn.1 TC1900000426.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003380.rn.1 TC1900000426.rn.1 RTA TC
          Rat Expression Set 230 1377302_a_at - -
          Rat Expression Set 230 1377302_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806993 - -
          Rat Genome U34 Set rc_AA943552_g_at - -
          Rat Genome U34 Set rc_AA943552_at - -
          Rat Genome U34 Set rc_AI145027_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21612203 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364617 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline